Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199820.2 | NP_956114.2 | 2 - 3 | 332 | 77 |
XM_005156611.2 | XP_005156668.1 | 2 - 3 | 243 | 77 | |
GenBank mRNA | BC059196.1 | - | 2 - 3 | 344 | 77 |
BC066670.1 | - | 2 - 3 | 332 | 77 |