Assay Design



Gene Transcripts

Gene Symbol
nup35
Entrez Gene ID
337461
Gene Name
nucleoporin 35
Gene Aliases
fj68d11, wu:fj68d11, zgc:65979
Chromosome Location
Chr.9: 12259172 - 12269982 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200066.1 NP_956360.1 7 - 8 740 65
XM_009304508.3 XP_009302783.1 7 - 8 849 65
GenBank mRNA BC061958.1 - 7 - 8 740 65

View Full Product Details