Assay Design
Gene Transcripts
- Gene Symbol
- paqr5a
- Entrez Gene ID
-
393156
- Gene Name
- progestin and adipoQ receptor family member Va
- Gene Aliases
-
paqr5, zgc:56016
- Chromosome Location
-
Chr.18: 53433 - 59606 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200187.1 |
NP_956481.1 |
4 - 5 |
631 |
90 |
GenBank mRNA |
BC045864.1 |
- |
4 - 5 |
631 |
90 |
View Full Product Details