Assay Design



Gene Transcripts

Gene Symbol
strap
Entrez Gene ID
393274
Gene Name
serine/threonine kinase receptor associated protein
Gene Aliases
zgc:56677, zgc:77604
Chromosome Location
Chr.4: 16876832 - 16883051 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200304.1 NP_956598.1 7 - 8 842 63
GenBank mRNA BC049525.1 - 7 - 8 842 63
BC065428.1 - 7 - 8 712 63

View Full Product Details