Assay Design



Gene Transcripts

Gene Symbol
anp32b
Entrez Gene ID
322000
Gene Name
acidic (leucine-rich) nuclear phosphoprotein 32 family, member B
Gene Aliases
cb908, fa99a11, wu:fa28c10, wu:fa99a11, wu:fb40e12, zgc:77745
Chromosome Location
Chr.1: 26660356 - 26666083 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212603.2 NP_997768.2 2 - 3 372 83
XM_005159865.4 XP_005159922.1 2 - 3 1600 83
GenBank mRNA BC065856.1 - 2 - 3 84
CV118158.1 - 2 - 3 323 83

View Full Product Details