Assay Design
Gene Transcripts
- Gene Symbol
- ythdf1
- Entrez Gene ID
-
327606
- Gene Name
- YTH N(6)-methyladenosine RNA binding protein 1
- Gene Aliases
-
fb49c02, wu:fb49c02, wu:fi19h06, wu:fj87f09, wu:fp06c11, zgc:55657
- Chromosome Location
-
Chr.8: 352151 - 353164 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_212713.1 |
NP_997878.1 |
4 - 5 |
2108 |
62 |
GenBank mRNA |
BC046885.1 |
- |
4 - 5 |
2108 |
62 |
View Full Product Details