Assay Design
Gene Transcripts
- Gene Symbol
- psmg2
- Entrez Gene ID
-
573107
- Gene Name
- proteasome (prosome, macropain) assembly chaperone 2
- Gene Aliases
-
tnfsf5ip1, wu:fb51b11, zgc:73278
- Chromosome Location
-
Chr.19: 12350110 - 12360121 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213032.1 |
NP_998197.1 |
1 - 2 |
137 |
71 |
GenBank mRNA |
BC059607.1 |
- |
1 - 2 |
137 |
71 |
View Full Product Details