Assay Design



Gene Transcripts

Gene Symbol
cdk5rap3
Entrez Gene ID
415195
Gene Name
CDK5 regulatory subunit associated protein 3
Gene Aliases
cdkrap3, fb33h04, fc09c05, id:ibd2920, wu:fb33h04, wu:fc09c05, zgc:86863
Chromosome Location
Chr.12: 268306 - 277620 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002105.1 NP_001002105.1 6 - 7 616 80
GenBank mRNA BC071504.1 - 6 - 7 616 80

View Full Product Details