Assay Design



Gene Transcripts

Gene Symbol
commd5
Entrez Gene ID
445159
Gene Name
COMM domain containing 5
Gene Aliases
zgc:101132
Chromosome Location
Chr.14: 10374530 - 10387363 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003553.2 NP_001003553.1 4 - 5 372 81
GenBank mRNA BC078284.1 - 4 - 5 377 81

View Full Product Details