Assay Design



Gene Transcripts

Gene Symbol
golim4b
Entrez Gene ID
541535
Gene Name
golgi integral membrane protein 4b
Gene Aliases
im:7146775, im:7156643, zgc:113641
Chromosome Location
Chr.2: 36701241 - 36721297 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014349.2 NP_001014371.2 1 - 2 431 105
XM_005171289.4 XP_005171346.1 1 - 2 512 105
GenBank mRNA BC091983.1 - 1 - 2 435 105

View Full Product Details