Assay Design



Gene Transcripts

Gene Symbol
cyp2ad3
Entrez Gene ID
569245
Gene Name
cytochrome P450, family 2, subfamily AD, polypeptide 3
Gene Aliases
cyp2j27, si:dkey-183n20.10
Chromosome Location
Chr.20: 25548425 - 25551676 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025554.2 NP_001020725.1 2 - 3 427 77
GenBank mRNA AY500819.1 - 2 - 3 347 77
BC085624.1 - 2 - 3 400 77
BC124073.1 - 2 - 3 394 77
BC134877.1 - 2 - 3 427 77

View Full Product Details