Assay Design



Gene Transcripts

Gene Symbol
slc4a4a
Entrez Gene ID
568631
Gene Name
solute carrier family 4 member 4a
Gene Aliases
id:ibd2520, nbce1, si:dkey-256m11.1, slc4a4
Chromosome Location
Chr.5: 45746922 - 45877126 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034984.1 NP_001030156.1 6 - 7 756 71
XM_009301743.3 XP_009300018.1 5 - 6 963 71
GenBank mRNA AY727858.1 - 6 - 7 756 71

View Full Product Details