Assay Design



Gene Transcripts

Gene Symbol
car15
Entrez Gene ID
568143
Gene Name
carbonic anhydrase 15
Gene Aliases
si:ch211-265m20.6
Chromosome Location
Chr.10: 175673 - 187935 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045139.1 NP_001038604.1 2 - 3 128 95
XM_005169221.4 XP_005169278.1 2 - 3 606 95

View Full Product Details