Assay Design



Gene Transcripts

Gene Symbol
pcsk5a
Entrez Gene ID
571235
Gene Name
proprotein convertase subtilisin/kexin type 5a
Gene Aliases
pc5.1, pcsk5.1, pcsk5/6a, si:dkey-267i17.1
Chromosome Location
Chr.5: 127333 - 215542 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021476634.1 XP_021332309.1 18 - 19 2599 104
GenBank mRNA DQ983232.1 - 18 - 19 2528 104

View Full Product Details