Assay Design



Gene Transcripts

Gene Symbol
mlpha
Entrez Gene ID
561905
Gene Name
melanophilin a
Gene Aliases
mlphb, si:ch211-186m12.3, zgc:154096
Chromosome Location
Chr.6: 57714 - 93409 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079679.3 NP_001073147.1 2 - 3 168 81
XM_005168711.4 XP_005168768.1 2 - 3 209 81
XM_005168712.4 XP_005168769.1 2 - 3 166 81
XM_005168713.4 XP_005168770.1 2 - 3 209 81
GenBank mRNA BC124539.1 - 1 - 2 120 81
DT063943.1 - 2 - 3 151 81

View Full Product Details