Assay Design



Gene Transcripts

Gene Symbol
git2b
Entrez Gene ID
567190
Gene Name
G protein-coupled receptor kinase interacting ArfGAP 2b
Gene Aliases
git2, zgc:158416
Chromosome Location
Chr.10: 50098 - 83574 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080188.2 NP_001073657.2 8 - 9 854 93
NM_001362634.1 NP_001349563.1 8 - 9 854 93
GenBank mRNA BC129227.1 - 8 - 9 853 93

View Full Product Details