Assay Design



Gene Transcripts

Gene Symbol
taar12g
Entrez Gene ID
794997
Gene Name
trace amine associated receptor 12g
Gene Aliases
si:dkey-7c18.20
Chromosome Location
Chr.20: 46068598 - 46069609 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082908.1 NP_001076377.1 1 - 1 852 102

View Full Product Details