Assay Design



Gene Transcripts

Gene Symbol
skib
Entrez Gene ID
30113
Gene Name
v-ski avian sarcoma viral oncogene homolog b
Gene Aliases
fc17g02, wu:fc17g02, wu:fk79g04
Chromosome Location
Chr.11: 396572 - 404947 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130938.1 NP_571013.1 3 - 4 1445 111
GenBank mRNA AF060118.1 - 3 - 4 1445 111
BC065946.1 - 3 - 4 1415 111

View Full Product Details