Assay Design



Gene Transcripts

Gene Symbol
men1
Entrez Gene ID
30130
Gene Name
multiple endocrine neoplasia I
Gene Aliases
wu:fc44c03, zgc:110510
Chromosome Location
Chr.7: 17691849 - 17712606 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130953.1 NP_571028.1 6 - 7 999 93
GenBank mRNA AF212919.1 - 6 - 7 999 93
AJ245952.1 - 6 - 7 988 93
BC095301.1 - 6 - 7 1058 93

View Full Product Details