Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001328376.1 | NP_001315305.1 | 1 - 2 | 622 | 69 |
NM_130963.2 | NP_571038.2 | 1 - 2 | 622 | 69 | |
GenBank mRNA | EH433465.1 | - | 1 - 2 | 651 | 69 |
EH579828.1 | - | 1 - 2 | 69 | ||
U47113.2 | - | 1 - 2 | 69 |