Assay Design
Gene Transcripts
- Gene Symbol
- elovl6
- Entrez Gene ID
-
317738
- Gene Name
- ELOVL fatty acid elongase 6
- Gene Aliases
-
cb618, lce, zgc:73089
- Chromosome Location
-
Chr.14: 9947 - 18628 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199532.1 |
NP_955826.1 |
2 - 3 |
346 |
128 |
GenBank mRNA |
BC059459.1 |
- |
2 - 3 |
346 |
128 |
View Full Product Details