Assay Design
Gene Transcripts
- Gene Symbol
- hist2h2l
- Entrez Gene ID
-
386920
- Gene Name
- histone 2, H2, like
- Gene Aliases
-
fj05g08, wu:fj05g08, zgc:73093
- Chromosome Location
-
Chr.10: 20090 - 20913 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200117.1 |
NP_956411.1 |
1 - 1 |
151 |
101 |
GenBank mRNA |
BC059463.1 |
- |
1 - 1 |
151 |
101 |
View Full Product Details