Assay Design



Gene Transcripts

Gene Symbol
ube2h
Entrez Gene ID
368425
Gene Name
ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast)
Gene Aliases
fc07a05, fi24c01, si:bz3c13.2, wu:fc07a05, wu:fi24c01
Chromosome Location
Chr.4: 15011341 - 15059876 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201489.1 NP_958897.1 5 - 6 416 94
GenBank mRNA BC049139.1 - 5 - 6 416 94
BC165255.1 - 5 - 6 330 94

View Full Product Details