Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201508.1 | NP_958916.1 | 9 - 10 | 1247 | 83 |
XM_005163231.4 | XP_005163288.1 | 9 - 10 | 1281 | 83 | |
XM_009297055.3 | XP_009295330.1 | 9 - 10 | 1292 | 83 | |
GenBank mRNA | BC047176.1 | - | 9 - 10 | 1247 | 83 |
BC165758.1 | - | 8 - 9 | 1235 | 83 |