Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203484.1 | NP_982351.1 | 26 - 27 | 3217 | 79 |
XM_021479576.1 | XP_021335251.1 | 26 - 27 | 3405 | 79 | |
XM_021479577.1 | XP_021335252.1 | 26 - 27 | 3405 | 79 | |
XM_021479578.1 | XP_021335253.1 | 26 - 27 | 3405 | 79 | |
XM_021479579.1 | XP_021335254.1 | 26 - 27 | 3405 | 79 | |
XM_021479580.1 | XP_021335255.1 | 26 - 27 | 3404 | 79 | |
XM_021479581.1 | XP_021335256.1 | 25 - 26 | 3329 | 79 | |
XM_021479582.1 | XP_021335257.1 | 24 - 25 | 3024 | 79 | |
XM_021479583.1 | XP_021335258.1 | 26 - 27 | 3405 | 79 | |
XM_021479584.1 | XP_021335259.1 | 25 - 26 | 3329 | 79 | |
GenBank mRNA | AY528224.1 | - | 26 - 27 | 3217 | 79 |
BC163679.1 | - | 24 - 25 | 3024 | 79 |