Assay Design



Gene Transcripts

Gene Symbol
creb3l3l
Entrez Gene ID
406853
Gene Name
cAMP responsive element binding protein 3-like 3 like
Gene Aliases
creb3l3, wu:fj05f07, zgc:66452
Chromosome Location
Chr.8: 131880 - 141311 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213532.2 NP_998697.2 9 - 10 1059 99
XM_005167154.3 XP_005167211.1 9 - 10 1214 99
GenBank mRNA BC055664.1 - 9 - 10 1107 99

View Full Product Details