Assay Design



Gene Transcripts

Gene Symbol
sult1st6
Entrez Gene ID
436872
Gene Name
sulfotransferase family 1, cytosolic sulfotransferase 6
Gene Aliases
sult1c3, zgc:92309
Chromosome Location
Chr.12: 15197857 - 15205064 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002599.1 NP_001002599.1 2 - 3 628 112
GenBank mRNA AY937249.1 - 2 - 3 314 112
BC075996.1 - 2 - 3 628 112

View Full Product Details