Assay Design



Gene Transcripts

Gene Symbol
glipr2l
Entrez Gene ID
449805
Gene Name
GLI pathogenesis-related 2, like
Gene Aliases
wu:fj53e10, zgc:103578
Chromosome Location
Chr.16: 27615037 - 27622183 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005978.1 NP_001005978.1 4 - 5 551 86
GenBank mRNA BC083432.1 - 4 - 5 551 86

View Full Product Details