Assay Design



Gene Transcripts

Gene Symbol
pacrg
Entrez Gene ID
492757
Gene Name
PARK2 co-regulated
Gene Aliases
zgc:101786
Chromosome Location
Chr.17: 27545214 - 27706746 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007398.2 NP_001007399.1 1 - 2 205 81
GenBank mRNA BC085440.1 - 1 - 2 206 81
BC165063.1 - 1 - 2 115 81

View Full Product Details