Assay Design



Gene Transcripts

Gene Symbol
ext1a
Entrez Gene ID
497279
Gene Name
exostosin glycosyltransferase 1a
Gene Aliases
fd07d06, wu:fd07d06
Chromosome Location
Chr.16: 48460873 - 48573130 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012368.2 NP_001012368.2 10 - 11 2577 60
GenBank mRNA AY734455.1 - 10 - 11 2008 60
BC133117.1 - 10 - 11 2577 60

View Full Product Details