Assay Design



Gene Transcripts

Gene Symbol
zcrb1
Entrez Gene ID
550251
Gene Name
zinc finger CCHC-type and RNA binding motif 1
Gene Aliases
si:ch211-155a11.4, zgc:110711
Chromosome Location
Chr.4: 13925287 - 13931515 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017589.2 NP_001017589.2 5 - 6 471 121
GenBank mRNA BC092992.1 - 5 - 6 446 121
EH551637.1 - 5 - 6 486 121
EH573039.1 - 3 - 4 656 121

View Full Product Details