Assay Design



Gene Transcripts

Gene Symbol
ndufb9
Entrez Gene ID
573021
Gene Name
NADH:ubiquinone oxidoreductase subunit B9
Gene Aliases
wu:fb10b03, zgc:110657
Chromosome Location
Chr.16: 74867 - 79201 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017880.1 NP_001017880.1 1 - 2 92 80
GenBank mRNA BC092968.1 - 1 - 2 92 80
BC154160.1 - 1 - 2 94 80

View Full Product Details