Assay Design



Gene Transcripts

Gene Symbol
gins2
Entrez Gene ID
554146
Gene Name
GINS complex subunit 2
Gene Aliases
si:dkey-265o13.3, zgc:112262
Chromosome Location
Chr.18: 30481030 - 30499456 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024439.1 NP_001019610.1 4 - 5 463 67
GenBank mRNA BC095711.1 - 4 - 5 463 67

View Full Product Details