Assay Design



Gene Transcripts

Gene Symbol
kcnk13b
Entrez Gene ID
556073
Gene Name
potassium channel, subfamily K, member 13b
Gene Aliases
kcnk13, si:ch211-173b9.3
Chromosome Location
Chr.20: 16703050 - 16709138 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025483.1 NP_001020654.1 1 - 2 337 128
GenBank mRNA BC163776.1 - 1 - 2 387 128
BC163778.1 - 1 - 2 387 128

View Full Product Details