Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037691.2 | NP_001032780.1 | 4 - 5 | 294 | 84 |
XM_021476902.1 | XP_021332577.1 | - | 1055 | 84 | |
GenBank mRNA | BC107825.1 | - | 4 - 5 | 294 | 84 |
EH441049.1 | - | 4 - 5 | 322 | 84 | |
EH549222.1 | - | 4 - 5 | 325 | 84 |