Assay Design



Gene Transcripts

Gene Symbol
olfcs1
Entrez Gene ID
564219
Gene Name
olfactory receptor C family, s1
Gene Aliases
si:dkey-188k17.1
Chromosome Location
Chr.18: 18502 - 23094 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045045.1 NP_001038510.1 1 - 2 176 65

View Full Product Details