Assay Design



Gene Transcripts

Gene Symbol
thbs3b
Entrez Gene ID
571317
Gene Name
thrombospondin 3b
Gene Aliases
si:dkey-81h8.2
Chromosome Location
Chr.19: 24416183 - 24443736 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045214.1 NP_001038679.1 15 - 16 1751 94
XM_005170062.4 XP_005170119.1 15 - 16 2162 94
GenBank mRNA BC163250.1 - 14 - 15 1709 94

View Full Product Details