Assay Design



Gene Transcripts

Gene Symbol
pdzrn4
Entrez Gene ID
798289
Gene Name
PDZ domain containing ring finger 4
Gene Aliases
si:dkey-240a12.3
Chromosome Location
Chr.4: 13811033 - 13861445 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082923.2 NP_001076392.1 1 - 2 64 75

View Full Product Details