Assay Design



Gene Transcripts

Gene Symbol
isy1
Entrez Gene ID
570205
Gene Name
ISY1 splicing factor homolog
Gene Aliases
im:7159238, wu:fc23h01, wu:fi21h07, zgc:136785
Chromosome Location
Chr.11: 474439 - 486306 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083563.2 NP_001077032.1 5 - 6 433 111
GenBank mRNA BC115265.1 - 5 - 6 207 111

View Full Product Details