Assay Design



Gene Transcripts

Gene Symbol
coq9
Entrez Gene ID
567716
Gene Name
coenzyme Q9 homolog (S. cerevisiae)
Gene Aliases
si:ch211-51l3.4
Chromosome Location
Chr.25: 14381405 - 14398265 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098746.1 NP_001092216.1 8 - 9 967 71
GenBank mRNA BC142917.1 - 8 - 9 967 71

View Full Product Details