Assay Design



Gene Transcripts

Gene Symbol
elf1
Entrez Gene ID
30396
Gene Name
E74-like ETS transcription factor 1
Gene Aliases
ef1, fb05d07, wu:fb05d07, zef1
Chromosome Location
Chr.14: 40745833 - 40850366 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131159.2 NP_571234.2 8 - 9 1483 93
XM_017358977.2 XP_017214466.1 8 - 9 1800 93
XM_017358978.2 XP_017214467.1 8 - 9 1595 93
XM_017358979.2 XP_017214468.1 8 - 9 1778 93
XM_017358980.2 XP_017214469.1 8 - 9 1797 93
GenBank mRNA U84615.1 - 8 - 9 1490 93

View Full Product Details