Assay Design



Gene Transcripts

Gene Symbol
psme1
Entrez Gene ID
30648
Gene Name
proteasome activator subunit 1
Gene Aliases
fk89d05, wu:fk89d05
Chromosome Location
Chr.24: 12945869 - 12954482 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131375.2 NP_571450.1 10 - 11 809 88
GenBank mRNA AF195052.1 - 10 - 11 810 88
BC075864.1 - 10 - 11 823 88

View Full Product Details