Assay Design
Gene Transcripts
- Gene Symbol
- epha4b
- Entrez Gene ID
-
64270
- Gene Name
- eph receptor A4b
- Gene Aliases
-
EphA3, epha4, id:ibd2782, rtk2
- Chromosome Location
-
Chr.2: 40294313 - 40514151 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_153658.1 |
NP_705944.1 |
15 - 16 |
2733 |
84 |
GenBank mRNA |
AF369381.1 |
- |
15 - 16 |
2733 |
84 |
View Full Product Details