Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178287.5 | NP_840072.3 | 3 - 4 | 248 | 81 |
GenBank mRNA | AY216582.1 | - | 3 - 4 | 159 | 81 |
BC092686.1 | - | 3 - 4 | 210 | 81 | |
BC152097.1 | - | 3 - 4 | 184 | 81 | |
BI472564.1 | - | 3 - 4 | 513 | 81 | |
CK395720.1 | - | 3 - 4 | 259 | 81 | |
DN892184.1 | - | 3 - 4 | 229 | 81 | |
EH445855.1 | - | 3 - 4 | 271 | 81 | |
EH564833.1 | - | 3 - 4 | 594 | 81 | |
EH593497.1 | - | 3 - 4 | 281 | 81 |