Assay Design



Gene Transcripts

Gene Symbol
slc7a4
Entrez Gene ID
796100
Gene Name
solute carrier family 7 member 4
Gene Aliases
si:busm1-266f07.3, si:dz266f07.3
Chromosome Location
Chr.8: 36500308 - 36515904 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194427.2 NP_919408.2 5 - 6 2176 62
GenBank mRNA BC134892.1 - 5 - 6 2176 62
BC171600.1 - 4 - 5 2115 62

View Full Product Details