Assay Design



Gene Transcripts

Gene Symbol
itm2ba
Entrez Gene ID
323632
Gene Name
integral membrane protein 2Ba
Gene Aliases
itm2b, wu:fc05f02, zgc:55770, zgc:85688
Chromosome Location
Chr.6: 10015525 - 10026822 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199646.1 NP_955940.1 4 - 5 637 69
GenBank mRNA BC044480.1 - 4 - 5 637 69
BC128819.1 - 4 - 5 613 69
BC164594.1 - 4 - 5 582 69

View Full Product Details