Assay Design



Gene Transcripts

Gene Symbol
edf1
Entrez Gene ID
793036
Gene Name
endothelial differentiation-related factor 1
Gene Aliases
zgc:73192, zgc:86829
Chromosome Location
Chr.5: 64397643 - 64406058 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200745.1 NP_957039.1 1 - 2 175 89
GenBank mRNA BC059541.1 - 1 - 2 175 89
BC071480.1 - 1 - 2 190 89
BC159201.1 - 1 - 2 163 89

View Full Product Details