Assay Design
Gene Transcripts
- Gene Symbol
- rab5ab
- Entrez Gene ID
-
393945
- Gene Name
- RAB5A, member RAS oncogene family, b
- Gene Aliases
-
Rab5a, rab5al, zgc:56644
- Chromosome Location
-
Chr.16: 49647436 - 49661406 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200970.1 |
NP_957264.1 |
2 - 3 |
257 |
110 |
GenBank mRNA |
BC049057.1 |
- |
2 - 3 |
257 |
110 |
View Full Product Details