Assay Design



Gene Transcripts

Gene Symbol
ppp6c
Entrez Gene ID
393980
Gene Name
protein phosphatase 6, catalytic subunit
Gene Aliases
zgc:56227
Chromosome Location
Chr.5: 31983106 - 31995401 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201005.1 NP_957299.1 2 - 3 244 129
GenBank mRNA BC047847.1 - 2 - 3 244 129
BC075751.1 - 2 - 3 208 129

View Full Product Details