Assay Design



Gene Transcripts

Gene Symbol
ndufs8a
Entrez Gene ID
406413
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8a
Gene Aliases
ndufs8, wu:fb30b07, zgc:65835, zgc:77840
Chromosome Location
Chr.1: 44814364 - 44823067 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213139.1 NP_998304.1 4 - 5 247 78
GenBank mRNA BC058299.1 - 4 - 5 247 78
BC065459.1 - 4 - 5 256 78

View Full Product Details